Search
Search

MAST1| Gene Symbol: MAST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761814.3 | XP_002761860.2 | 18 - 19 | 2254 | 58 | |
| XM_008987365.2 | XP_008985613.1 | 19 - 20 | 2169 | 58 | ||
| XM_017967528.1 | XP_017823017.1 | 18 - 19 | 2254 | 58 | ||
Set Membership: |
|