Search
Search

CYP17A1| Gene Symbol: CYP17A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267768.1 | NP_001254697.1 | 3 - 4 | 671 | 102 | |
| GenBank mRNA | AY746982.1 | - | 3 - 4 | 671 | 102 | |
Set Membership: |
|