Search
Search

ROCK1| Gene Symbol: ROCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979587.2 | XP_008977835.1 | 28 - 29 | 3420 | 98 | |
| XM_017963914.1 | XP_017819403.1 | 27 - 28 | 3197 | 98 | ||
Set Membership: |
|