Search
Search

LAYN| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754375.4 | XP_002754421.1 | 5 - 6 | 1414 | 114 | |
| XM_009006760.2 | XP_009005008.1 | 5 - 6 | 1096 | 114 | ||
| XM_009006761.2 | XP_009005009.1 | 3 - 4 | 223 | 114 | ||
Set Membership: |
|