Search
Search

NCKIPSD| Gene Symbol: NCKIPSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758318.3 | XP_002758364.1 | 5 - 6 | 1182 | 99 | |
| XM_008981600.2 | XP_008979848.1 | 6 - 7 | 1364 | 99 | ||
| XM_008981601.1 | XP_008979849.1 | 5 - 6 | 1189 | 99 | ||
Set Membership: |
|