Search
Search

IFT140| Gene Symbol: IFT140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008894.2 | XP_009007142.1 | 3 - 4 | 1004 | 56 | |
| XM_009008895.2 | XP_009007143.1 | 3 - 4 | 619 | 56 | ||
| XM_017978611.1 | XP_017834100.1 | 3 - 4 | 1007 | 56 | ||
Set Membership: |
|