Search
Search

TOP2A| Gene Symbol: TOP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748560.2 | XP_002748606.1 | 32 - 33 | 4304 | 101 | |
| XM_017972718.1 | XP_017828207.1 | 32 - 33 | 4709 | 101 | ||
Set Membership: |
|