Search
Search

AATF| Gene Symbol: AATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748315.3 | XP_002748361.2 | 2 - 3 | 532 | 74 | |
| XM_008997005.2 | XP_008995253.1 | 2 - 3 | 532 | 74 | ||
| XM_008997006.1 | XP_008995254.1 | 2 - 3 | 532 | 74 | ||
Set Membership: |
|