Search
Search

NOP9| Gene Symbol: NOP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005833.1 | XP_009004081.1 | 8 - 9 | 1800 | 84 | |
| XM_009005835.1 | XP_009004083.1 | 8 - 9 | 1758 | 84 | ||
| XM_009005836.1 | XP_009004084.1 | 8 - 9 | 1589 | 84 | ||
Set Membership: |
|