Search
Search

ORC5| Gene Symbol: ORC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751686.1 | XP_002751732.1 | 8 - 9 | 960 | 84 | |
| XM_017975430.1 | XP_017830919.1 | 8 - 9 | 960 | 84 | ||
| XM_017975431.1 | XP_017830920.1 | 7 - 8 | 871 | 84 | ||
Set Membership: |
|