Search
Search

LTBR| Gene Symbol: LTBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003462.1 | XP_009001710.1 | 3 - 4 | 646 | 97 | |
| XM_009003463.1 | XP_009001711.1 | 3 - 4 | 646 | 97 | ||
| XM_017975878.1 | XP_017831367.1 | 3 - 4 | 607 | 97 | ||
Set Membership: |
|