Search
Search

PDCD10| Gene Symbol: PDCD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759415.4 | XP_002759461.1 | 4 - 5 | 504 | 74 | |
| XM_008983515.2 | XP_008981763.1 | 5 - 6 | 565 | 74 | ||
| XM_008983516.1 | XP_008981764.1 | 4 - 5 | 395 | 74 | ||
| XM_008983517.2 | XP_008981765.1 | 4 - 5 | 612 | 74 | ||
Set Membership: |
|