Search
Search

FLOT2| Gene Symbol: FLOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748231.4 | XP_002748277.3 | 9 - 10 | 1953 | 92 | |
| XM_008997161.2 | XP_008995409.1 | 10 - 11 | 2060 | 92 | ||
| XM_008997162.2 | XP_008995410.1 | 10 - 11 | 2060 | 92 | ||
| XM_008997163.2 | XP_008995411.1 | 9 - 10 | 1253 | 92 | ||
| XM_008997165.2 | XP_008995413.1 | 9 - 10 | 1138 | 92 | ||
| XM_017972566.1 | XP_017828055.1 | 8 - 9 | 1055 | 92 | ||
Set Membership: |
|