Search
Search

RAVER2| Gene Symbol: RAVER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001401.2 | XP_008999649.1 | 9 - 10 | 1562 | 66 | |
| XM_009001402.2 | XP_008999650.1 | 9 - 10 | 1559 | 66 | ||
| XM_009001403.2 | XP_008999651.1 | 9 - 10 | 1523 | 66 | ||
Set Membership: |
|