Search
Search

MOK| Gene Symbol: MOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006574.1 | XP_009004822.1 | 12 | 1670 | 87 | |
| XM_009006577.2 | XP_009004825.1 | 12 | 1462 | 87 | ||
| XM_009006578.1 | XP_009004826.1 | 11 | 1469 | 87 | ||
| XM_017977338.1 | XP_017832827.1 | 13 | 1838 | 87 | ||
| XM_017977339.1 | XP_017832828.1 | 9 | 1657 | 87 | ||
| XM_017977340.1 | XP_017832829.1 | 9 | 1354 | 87 | ||
Set Membership: |
|