Search
Search

CD48| Gene Symbol: CD48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760232.4 | XP_002760278.2 | 2 - 3 | 199 | 75 | |
| XM_008984745.1 | XP_008982993.1 | 2 - 3 | 241 | 75 | ||
| XM_017966404.1 | XP_017821893.1 | 2 - 3 | 412 | 75 | ||
Set Membership: |
|