Search
Search

AKAP8L| Gene Symbol: AKAP8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761844.4 | XP_002761890.1 | 9 - 10 | 1382 | 60 | |
| XM_008987439.2 | XP_008985687.1 | 10 - 11 | 1256 | 60 | ||
| XM_017967424.1 | XP_017822913.1 | 9 - 10 | 1310 | 60 | ||
| XM_017967425.1 | XP_017822914.1 | 9 - 10 | 1310 | 60 | ||
| XM_017967426.1 | XP_017822915.1 | 9 - 10 | 1310 | 60 | ||
| XM_017967427.1 | XP_017822916.1 | 9 - 10 | 1310 | 60 | ||
Set Membership: |
|