Search
Search

DOCK1| Gene Symbol: DOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008978903.2 | XP_008977151.1 | 43 - 44 | 4593 | 67 | |
| XM_017963271.1 | XP_017818760.1 | 43 - 44 | 4530 | 67 | ||
Set Membership: |
|