Search
Search

CYB5R4| Gene Symbol: CYB5R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746774.3 | XP_002746820.1 | 3 - 4 | 461 | 83 | |
| XM_008994727.2 | XP_008992975.1 | 3 - 4 | 463 | 83 | ||
| XM_008994728.2 | XP_008992976.1 | 3 - 4 | 463 | 83 | ||
Set Membership: |
|