Search
Search

TAC4| Gene Symbol: TAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748441.3 | XP_002748487.1 | 1 - 2 | 389 | 89 | |
| XM_008997391.2 | XP_008995639.1 | 1 - 2 | 386 | 89 | ||
| XM_008997392.2 | XP_008995640.1 | 1 - 2 | 387 | 89 | ||
| XM_008997393.2 | XP_008995641.1 | 1 - 2 | 356 | 89 | ||