Search
Search

C1H9orf43| Gene Symbol: C1H9orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975720.1 | XP_017831209.1 | 2 - 3 | 2088 | 78 | |
| XM_017975721.1 | XP_017831210.1 | 1 - 2 | 2456 | 78 | ||
| XM_017975722.1 | XP_017831211.1 | 1 - 2 | 2455 | 78 | ||
| XM_017975724.1 | XP_017831213.1 | 1 - 2 | 2455 | 78 | ||
| XM_017975725.1 | XP_017831214.1 | 1 - 2 | 2459 | 78 | ||
| XM_017975727.1 | XP_017831216.1 | 1 - 2 | 2489 | 78 | ||
Set Membership: |
|