Search
Search

CAJA-DMB| Gene Symbol: CAJA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746418.3 | XP_002746464.1 | 5 - 6 | 1739 | 85 | |
| GenBank mRNA | EF215139.1 | - | 3 - 4 | 375 | 85 | |
Set Membership: |
|