Search
Search

FBXO31| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761240.2 | XP_002761286.1 | 4 - 5 | 715 | 76 | |
| XM_008986333.2 | XP_008984581.1 | 4 - 5 | 715 | 76 | ||
| XM_017966813.1 | XP_017822302.1 | 4 - 5 | 715 | 76 | ||
Set Membership: |
|