Search
Search

PTPRR| Gene Symbol: PTPRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752758.3 | XP_002752804.1 | 12 - 13 | 2109 | 63 | |
| XM_009004212.2 | XP_009002460.1 | 11 - 12 | 1576 | 63 | ||
Set Membership: |
|