Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cj06018375_m1
          See other LRRFIP1 GE Assays ›
          Gene Symbol:
          LRRFIP1
          Gene Name:
          LRR binding FLII interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 154209486 - 154277476 on Build Callithrix_jacchus_cj1700_1.1
          Species:
          Callithrix jacchus
          Species Specific ID (Flybase ID):
          -
          Assay ID Cj06018375_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LRRFIP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002749962.4 XP_002750008.4 15 - 16 1140 96
          XM_017973820.1 XP_017829309.1 16 - 17 1243 96
          XM_017973821.1 XP_017829310.1 16 - 17 1237 96
          XM_017973822.1 XP_017829311.1 16 - 17 1219 96
          XM_017973823.1 XP_017829312.1 16 - 17 1210 96
          XM_017973824.1 XP_017829313.1 17 - 18 1283 96
          XM_017973825.1 XP_017829314.1 15 - 16 1159 96
          XM_017973826.1 XP_017829315.1 15 - 16 1141 96
          XM_017973827.1 XP_017829316.1 14 - 15 1095 96
          XM_017973828.1 XP_017829317.1 17 - 18 1283 96
          XM_017973829.1 XP_017829318.1 13 - 14 1056 96
          XM_017973830.1 XP_017829319.1 13 - 14 1029 96
          XM_017973831.1 XP_017829320.1 16 - 17 1210 96
          XM_017973832.1 XP_017829321.1 13 - 14 2666 96
          XM_017973833.1 XP_017829322.1 12 - 13 990 96
          XM_017973834.1 XP_017829323.1 12 - 13 951 96
          XM_017973835.1 XP_017829324.1 11 - 12 930 96
          XM_017973836.1 XP_017829325.1 11 - 12 918 96
          XM_017973837.1 XP_017829326.1 11 - 12 912 96
          XM_017973838.1 XP_017829327.1 11 - 12 879 96
          XM_017973839.1 XP_017829328.1 10 - 11 852 96
          XM_017973840.1 XP_017829329.1 12 - 13 951 96
          XM_017973841.1 XP_017829330.1 6 - 7 694 96
          XM_017973842.1 XP_017829331.1 10 - 11 852 96
          XM_017973843.1 XP_017829332.1 5 - 6 622 96
          XM_017973844.1 XP_017829333.1 9 - 10 780 96
          XM_017973845.1 XP_017829334.1 5 - 6 622 96
          XM_017973846.1 XP_017829335.1 17 - 18 1283 96
          XM_017973847.1 XP_017829336.1 16 - 17 1237 96
          XM_017973848.1 XP_017829337.1 14 - 15 1135 96
          XM_017973849.1 XP_017829338.1 17 - 18 1283 96
          XM_017973850.1 XP_017829339.1 5 - 6 622 96

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline