Search
Search

CDK14| Gene Symbol: CDK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002498.1 | XP_009000746.1 | 13 - 14 | 1542 | 93 | |
| XM_009002499.2 | XP_009000747.1 | 13 - 14 | 1435 | 93 | ||
| XM_009002500.1 | XP_009000748.1 | 12 - 13 | 1339 | 93 | ||
| XM_009002501.2 | XP_009000749.1 | 12 - 13 | 1612 | 93 | ||
| XM_017975361.1 | XP_017830850.1 | 13 - 14 | 1542 | 93 | ||
| XM_017975362.1 | XP_017830851.1 | 13 - 14 | 1542 | 93 | ||
| XM_017975363.1 | XP_017830852.1 | 13 - 14 | 1542 | 93 | ||
Set Membership: |
|