Search
Search

MPP2| Gene Symbol: MPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996714.2 | XP_008994962.1 | 5 - 6 | 752 | 60 | |
| XM_017972382.1 | XP_017827871.1 | 5 - 6 | 809 | 60 | ||
| XM_017972383.1 | XP_017827872.1 | 5 - 6 | 684 | 60 | ||
Set Membership: |
|