Search
Search

ANO2| Gene Symbol: ANO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017975874.1 | XP_017831363.1 | 24 - 25 | 3739 | 59 | |
| XM_017975875.1 | XP_017831364.1 | 23 - 24 | 4707 | 59 | ||
| XM_017975876.1 | XP_017831365.1 | 23 - 24 | 3167 | 59 | ||
Set Membership: |
|