Search
Search

RTBDN| Gene Symbol: RTBDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987369.2 | XP_008985617.1 | 2 - 3 | 586 | 67 | |
| XM_008987370.2 | XP_008985618.1 | 2 - 3 | 1168 | 67 | ||
| XM_017967529.1 | XP_017823018.1 | 2 - 3 | 1170 | 67 | ||
Set Membership: |
|