Search
Search

CLEC7A| Gene Symbol: CLEC7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752160.3 | XP_002752206.1 | 4 - 5 | 667 | 102 | |
| XM_002752162.3 | XP_002752208.1 | 3 - 4 | 529 | 102 | ||
Set Membership: |
|