Search
Search

DNMT3B| Gene Symbol: DNMT3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971844.1 | XP_017827333.1 | 20 - 21 | 2642 | 102 | |
| XM_017971848.1 | XP_017827337.1 | 21 - 22 | 2703 | 102 | ||
Set Membership: |
|