Search
Search

WNT10B| Gene Symbol: WNT10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752421.4 | XP_002752467.1 | 5 - 6 | 1444 | 67 | |
| XM_009003713.2 | XP_009001961.1 | 4 - 5 | 1761 | 67 | ||
| XM_009003717.2 | XP_009001965.1 | 5 - 6 | 1437 | 67 | ||
| XM_009003718.1 | XP_009001966.1 | 3 - 4 | 1648 | 67 | ||
| XM_017976027.1 | XP_017831516.1 | 2 - 3 | 3601 | 67 | ||
Set Membership: |
|