Search
Search

PFKFB3| Gene Symbol: PFKFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750028.4 | XP_002750074.1 | 6 - 7 | 879 | 70 | |
| XM_008999885.2 | XP_008998133.1 | 6 - 7 | 879 | 70 | ||
| XM_008999890.1 | XP_008998138.1 | 6 - 7 | 639 | 70 | ||
| XM_017974032.1 | XP_017829521.1 | 6 - 7 | 709 | 70 | ||
| XM_017974033.1 | XP_017829522.1 | 6 - 7 | 705 | 70 | ||
| XM_017974034.1 | XP_017829523.1 | 6 - 7 | 618 | 70 | ||
| XM_017974035.1 | XP_017829524.1 | 6 - 7 | 879 | 70 | ||
| XM_017974036.1 | XP_017829525.1 | 6 - 7 | 698 | 70 | ||
| XM_017974037.1 | XP_017829526.1 | 6 - 7 | 703 | 70 | ||
| XM_017974038.1 | XP_017829527.1 | 6 - 7 | 703 | 70 | ||
| XM_017974039.1 | XP_017829528.1 | 6 - 7 | 639 | 70 | ||
| XM_017974040.1 | XP_017829529.1 | 6 - 7 | 533 | 70 | ||
| XM_017974041.1 | XP_017829530.1 | 6 - 7 | 879 | 70 | ||
| XM_017974043.1 | XP_017829532.1 | 6 - 7 | 488 | 70 | ||
| XM_017974044.1 | XP_017829533.1 | 6 - 7 | 639 | 70 | ||
| XM_017974045.1 | XP_017829534.1 | 6 - 7 | 704 | 70 | ||
| XM_017974046.1 | XP_017829535.1 | 6 - 7 | 879 | 70 | ||
Set Membership: |
|