Search
Search

ZNF436| Gene Symbol: ZNF436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000610.2 | XP_008998858.1 | 2 - 3 | 535 | 70 | |
| XM_009000612.2 | XP_008998860.1 | 3 - 4 | 2677 | 70 | ||
| XM_009000613.2 | XP_008998861.1 | 2 - 3 | 2622 | 70 | ||
Set Membership: |
|