Search
Search

MKKS| Gene Symbol: MKKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747431.3 | XP_002747477.1 | 3 - 4 | 1971 | 81 | |
| XM_017971958.1 | XP_017827447.1 | 4 - 5 | 1821 | 81 | ||
| XM_017971959.1 | XP_017827448.1 | 4 - 5 | 2299 | 81 | ||
| XM_017971960.1 | XP_017827449.1 | 3 - 4 | 1654 | 81 | ||
Set Membership: |
|