Search
Search

SPATC1| Gene Symbol: SPATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965520.1 | XP_017821009.1 | 1 - 2 | 306 | 85 | |
| XM_017965521.1 | XP_017821010.1 | 1 - 2 | 306 | 85 | ||
Set Membership: |
|