Search
Search

PRSS53| Gene Symbol: PRSS53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756092.3 | XP_002756138.1 | 4 - 5 | 919 | 65 | |
| XM_009009466.2 | XP_009007714.1 | 2 - 3 | 1911 | 65 | ||
Set Membership: |
|