Search
Search

ZNF695| Gene Symbol: ZNF695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985645.2 | XP_008983893.1 | 4 - 5 | 479 | 82 | |
| XM_008985646.2 | XP_008983894.1 | 4 - 5 | 464 | 82 | ||
| XM_008985648.2 | XP_008983896.1 | 3 - 4 | 399 | 82 | ||
| XM_017966657.1 | XP_017822146.1 | 3 - 4 | 1492 | 82 | ||
Set Membership: |
|