Search
Search

Crybb3| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173070.1 | NP_001166541.1 | 2 - 3 | 145 | 83 | |
| GenBank mRNA | EB370163.1 | - | 2 - 3 | 132 | 83 | |
| EF457997.1 | - | 2 - 3 | 145 | 83 | ||
Set Membership: |
|