Search
Search

Crybb1| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173078.1 | NP_001166549.1 | 4 - 5 | 465 | 69 | |
| XM_013141520.2 | XP_012996974.1 | 3 - 4 | 455 | 69 | ||
| GenBank mRNA | EB369451.1 | - | 4 - 5 | 463 | 69 | |
| EF666993.1 | - | 4 - 5 | 465 | 69 | ||
Set Membership: |
|