Search
Search

CG7470| Gene Symbol: CG7470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259948.2 | NP_001246877.1 | 9 - 10 | 1706 | 109 | |
| NM_141118.5 | NP_649375.1 | 9 - 10 | 1706 | 109 | ||
| GenBank mRNA | AY069110.1 | - | 9 - 10 | 1682 | 109 | |
Set Membership: |
|