Search
Search

en| Gene Symbol: en | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078976.4 | NP_523700.2 | 1 - 2 | 1524 | 67 | |
| NM_165841.2 | NP_725059.1 | 1 - 2 | 1851 | 67 | ||
| GenBank mRNA | AY069448.1 | - | 1 - 2 | 1504 | 67 | |
Set Membership: |
|