Search
Search

MCPH1| Gene Symbol: MCPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169651.2 | NP_001163122.1 | 5 - 6 | 1020 | 59 | |
| NM_001169652.2 | NP_001163123.1 | 6 - 7 | 1231 | 59 | ||
| NM_136867.3 | NP_610711.1 | 6 - 7 | 1231 | 59 | ||
| NM_165852.2 | NP_725088.1 | 5 - 6 | 1020 | 59 | ||
| GenBank mRNA | AY122187.1 | - | 6 - 7 | 1195 | 59 | |
| BT088945.1 | - | 5 - 6 | 1027 | 59 | ||
| EF587234.1 | - | 5 - 6 | 1020 | 59 | ||
Set Membership: |
|