Search
Search

fdl| Gene Symbol: fdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299421.1 | NP_001286350.1 | 3 - 4 | 1218 | 81 | |
| NM_001299422.1 | NP_001286351.1 | 3 - 4 | 852 | 81 | ||
| NM_001370052.1 | NP_001356962.1 | 3 - 4 | 1218 | 81 | ||
| NM_165908.2 | NP_725178.2 | 3 - 4 | 748 | 81 | ||
| NM_165909.3 | NP_725179.1 | 3 - 4 | 1218 | 81 | ||
| GenBank mRNA | AF323977.1 | - | 3 - 4 | 1221 | 81 | |
Set Membership: |
|