Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01793249_g1
See other GALPHAQ GE Assays ›
Gene Symbol:
Galphaq
Gene Name:
G protein alpha q subunit
Gene Aliases:
CG17759, DGalpha[[q]], DGalphaq, DGq, DGqalpha, Dgq, DmGalpha[[q]], Dmel\CG17759, Dmel_CG17759, Dromel_CG17759_galpha49b_mORF, G alpha 49B, G?q, GNAQ/11/14, G[[a]]q, G[[alphaq]], G[[q]], G[[q]]alpha, G[[qalpha]], G[q], Galpha, Galpha49B, Galpha49b, Galpha[[q]], Gaq, Gq, Gqa-3, Gqalpha, Gqalpha[[e]], Gqalphae, Gqalpha{e}, dG[[alphaq]], dG[[q]]alpha, dGaalpha, dGq, dGqa-49b, dGqalpha, dGqalpha-3, dGqalpha-49b, dgq, dgqalpha, galpha49B, sast
Chromosome Location:
Chr.2R: 12612740 - 12623197 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004435
Assay ID
Dm01793249_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Galphaq
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_165924.2
NP_725196.1
8 - 9
1409
75
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Drosophila melanogaster
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
mucosal immune response
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
activation of phospholipase C activity
phototransduction
sensory perception of smell
flight behavior
rhodopsin mediated signaling pathway
metarhodopsin inactivation
axon midline choice point recognition
stabilization of membrane potential
thermotaxis
negative regulation of compound eye retinal cell programmed cell death
phospholipase C-activating dopamine receptor signaling pathway
neuron cellular homeostasis
cellular response to carbon dioxide
regulation of neuromuscular synaptic transmission
G-protein coupled receptor binding
GTPase activity
guanyl nucleotide binding
G-protein beta/gamma-subunit complex binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline