Search
Search

Dgkepsilon| Gene Symbol: Dgkepsilon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299433.1 | NP_001286362.1 | 3 - 4 | 1600 | 78 | |
| NM_165949.2 | NP_725228.1 | 3 - 4 | 1684 | 78 | ||
| GenBank mRNA | AY050241.1 | - | 3 - 4 | 1567 | 78 | |
Set Membership: |
|