Search
Search

Dh31-R| Gene Symbol: Dh31-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274021.2 | NP_001260950.1 | 2 - 3 | 855 | 75 | |
| NM_001274022.2 | NP_001260951.1 | 2 - 3 | 855 | 75 | ||
| NM_165979.3 | NP_725278.1 | 2 - 3 | 855 | 75 | ||
Set Membership: |
|