Search
Search

Eip75B| Gene Symbol: Eip75B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259892.1 | NP_001246821.1 | 3 - 4 | 2674 | 164 | |
| NM_001259893.2 | NP_001246822.1 | 3 - 4 | 2656 | 164 | ||
| NM_168755.1 | NP_730321.1 | 1 - 2 | 1361 | 164 | ||
Set Membership: |
|