Search
Search

verm| Gene Symbol: verm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169998.3 | NP_001163469.1 | 6 - 7 | 628 | 55 | |
| NM_168808.4 | NP_730441.1 | 5 - 6 | 556 | 55 | ||
| NM_168809.4 | NP_730442.2 | 5 - 6 | 538 | 55 | ||
| GenBank mRNA | BT016093.1 | - | 5 - 6 | 527 | 55 | |
Set Membership: |
|