Search
Search

SREBP| Gene Symbol: SREBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275135.1 | NP_001262064.1 | 4 - 5 | 2626 | 92 | |
| NM_079442.3 | NP_524166.2 | 3 - 4 | 2524 | 92 | ||
| NM_168815.2 | NP_730449.1 | 4 - 5 | 2547 | 92 | ||
| NM_168816.2 | NP_730450.1 | 4 - 5 | 2657 | 92 | ||
| GenBank mRNA | BT010285.1 | - | 4 - 5 | 2538 | 92 | |
Set Membership: |
|